Order Entry
Italy
Orders LinkContactUsLinkComponent
D-(+)-Glucose monohydrate

D-(+)-Glucose monohydrate

Glass bottle for solids, 500 g
Catalog # A11090.36
D-(+)-Glucose monohydrate
Catalog # A11090.36
CAS Number:  14431-43-7
D-(+)-Glucose monohydrate
D-(+)-Glucose monohydrate

Glass bottle for solids, 500 g

CAS Number:  14431-43-7
Price
25,60 €
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 14431-43-7 Molecular Formula: C6H12O6.H2O Formula Weight: 198.18 (180.16anhy) Physical Form: Cryst powder Appearance: White Melting Point: ca 83° MDL No.: MFCD00149450

Synonyms:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS Number:14431-43-7
  • Formula:C₆H₁₂O₆·H₂O
  • MDL Number:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Melting Pt:∼ 83 °C
  • Storage Temperature:Ambient
  • EINECS:200-075-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.