Order Entry
Italy
Orders LinkContactUsLinkComponent
Slide wallets, FiloSlide™

Slide wallets, FiloSlide™

Six wallets and index strips, Transparent, 260×380 mm
Catalog # 720-2346
Supplier:   CELLPATH
Slide wallets, FiloSlide™
Catalog # 720-2346
Supplier:  CELLPATH
Slide wallets, FiloSlide™
Slide wallets, FiloSlide™

Six wallets and index strips, Transparent, 260×380 mm

Price
31,90 €
Log in for availability
1 * 10 pezzi
Product Details & Documents

These slide folders feature 21 compartments for 76×26, 50×76 and 76×76 mm slides.

  • Hanging system
  • Storage in conventional filing cabinets
Specifications
  • W×D:
    260×380 mm
  • Colour:
    Transparent
  • Description:
    Six wallets and index strips
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.