Order Entry
Italy
ContactUsLinkComponent
Hand Held Refractometer MASTER-10M MASTER-10 series is suitable for particularly low concentrations of Brix 10% or less, such as tomato juice, vegetables, soups, cleaning solutions and cutting oil. This model does not have automatic temperature compe 1 * 1 items
Catalog # 635-0696
Nome del fornitore:  Atago
undefined
Hand Held Refractometer MASTER-10M MASTER-10 series is suitable for particularly low concentrations of Brix 10% or less, such as tomato juice, vegetables, soups, cleaning solutions and cutting oil. This model does not have automatic temperature compe 1 * 1 items
Catalog # 635-0696
Nome del fornitore:  Atago
Frequently Bought Together