Order Entry
Italy
Orders LinkContactUsLinkComponent
Eggertz tubes for comparators

Eggertz tubes for comparators

Eggertz tubes, 50 ml with 0,1 ml division for comparators
Catalog # WITG2.149.050
Supplier:   witeg Labortechnik
Eggertz tubes for comparators
Catalog # WITG2.149.050
Supplier:  witeg Labortechnik
Eggertz tubes for comparators
Eggertz tubes for comparators

Eggertz tubes, 50 ml with 0,1 ml division for comparators

Price
53,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Eggertz tubes, 50 ml with 0,1 ml division for comparators
  • Seq:
    02