Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Tube collar used as a weight for the aspiration tube or as a clamp spacer for the dispensing tube, 4 − 6 mm
Catalog # 710-1041
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1041
Accessories for DOSE IT
Accessories for DOSE IT
Tube collar used as a weight for the aspiration tube or as a clamp spacer for the dispensing tube, 4 − 6 mm
Price
77,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Tube collar used as a weight for the aspiration tube or as a clamp spacer for the dispensing tube
  • Ø int.:
    4 − 6 mm