SIELC PRIMESEP 100 COLUMN 150MM, 4.6 MM, 1 * 1 items
Catalog # SIEL10046.150.0510
Supplier: SIELC Technologies
SIELC PRIMESEP 100 COLUMN 150MM, 4.6 MM, 1 * 1 items
Catalog # SIEL10046.150.0510
Supplier: SIELC Technologies
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Columns, PD MidiTrap™ G-25
Supplier: Cytiva
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Crown ether columns, CROWNPAK®
Supplier: CHIRAL TECHNOLOGIES
Avantor® ACE® Silica, HPLC Columns, 5 µm
Supplier: Avantor