Order Entry
Italy
ContactUsLinkComponent
PowerCHO 1 chemically defined, serum-free CHO medium, with HEPES and Pluronic, without L-glutamine, phenol red, hypoxanthine, and thymidine 1 * 1 l
Catalog # LONZ12-770Q
Supplier:  LONZA
PowerCHO 1 chemically defined, serum-free CHO medium, with HEPES and Pluronic, without L-glutamine, phenol red, hypoxanthine, and thymidine 1 * 1 l
Catalog # LONZ12-770Q
Supplier:  LONZA
Frequently Bought Together