Order Entry
Italy
ContactUsLinkComponent
Areometer, DIN12791 L50 series, measuring range: 0.80-0.85:0.0005g/cm , without thermometer, reference temperature:20°C, length:335m m, suitable for government verification, with verification certificate. 1 * 1 items
Catalog # AMARH806208/VC
Supplier:  Amarell
Areometer, DIN12791 L50 series, measuring range: 0.80-0.85:0.0005g/cm , without thermometer, reference temperature:20°C, length:335m m, suitable for government verification, with verification certificate. 1 * 1 items
Catalog # AMARH806208/VC
Supplier:  Amarell
Frequently Bought Together