Digital Solutions
Applications
Support
About
Italy
Orders Link
ContactUsLinkComponent
Racks
Cryogenic Box Racks
Storing systems/racks and roller bases for ARPEGE
Storing systems/racks and roller bases for ARPEGE
Set of 6 racks, 9 levels for 2 ml vials, ARPEGE 170
AIRLACCRACK303
:
Air Liquide
Storing systems/racks and roller bases for ARPEGE
AIRLACCRACK303
:
Air Liquide
Storing systems/racks and roller bases for ARPEGE
Set of 6 racks, 9 levels for 2 ml vials, ARPEGE 170
1 190,00 €
1 * 1 SET
-
+
For:
ARPEGE 170
Description:
Set of 6 racks, 9 levels for 2 ml vials
Seq:
45