Triethylammonium bicarbonate buffer, 1 M, volatile buffer for HPLC, Fluka™
Triethylammonium bicarbonate buffer, 1 M, 8,4 - 8,6, Glass bottle, 500 mL
Catalog # 17902-500ML
Supplier: Honeywell Chemicals
Triethylammonium bicarbonate buffer, 1 M, volatile buffer for HPLC, Fluka™
Catalog # 17902-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Triethylammonium bicarbonate buffers are volatile buffer for HPLC.
- Non-volatile matter: max. 0,05 %
- pH (20 °C): 8,4 to 8,6
- Absorbance at 260 nm: Max. 0,04
- Absorbance at 280 nm: Max. 0,04
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:Triethylammonium bicarbonate buffer, 1 M
- pH value:8,4 - 8,6
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



