Order Entry
Italy
Orders LinkContactUsLinkComponent
Hydrogen peroxide 30%, Supelco®

Hydrogen peroxide 30%, Supelco®

Glass bottle, 500 mL
Codice catalogo 95321-500ML
Corrosive
Irritant
Nome del fornitore:   Merck
Hydrogen peroxide 30%, Supelco®
Codice catalogo 95321-500ML
Nome del fornitore:  Merck
Numero CAS:  7722-84-1
Hydrogen peroxide 30%, Supelco®
Hydrogen peroxide 30%, Supelco®

Glass bottle, 500 mL

Numero CAS:  7722-84-1
Price
196,00 €
Log in for availability
1 * 500 mL
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. Ulteriori informazioni
Product Details & Documents
  • CAS Number:7722-84-1
  • Formula:H₂O₂
  • MDL Number:MFCD00011333
  • Molecular Weight:34.01 g/mol
  • Boiling Pt:∼107 °C (1013 hPa)
  • Melting Pt:< 0 °C
  • Density:1.11 g/cm³ (20 °C)
  • ADR:5.1,II
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 1.11 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-765-0
  • UN:2014

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.