Order Entry
Italy
Orders LinkContactUsLinkComponent
2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®

2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®

Glass bottle for solids, 1 g
Catalog # 91928-1G
Irritant
Supplier:   Merck
2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®
Catalog # 91928-1G
Supplier:  Merck
CAS Number:  207-556-5
2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®
2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®

Glass bottle for solids, 1 g

CAS Number:  207-556-5
Price
74,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2-Acetylphloroglucinol monohydrate, THAP monohydrate, Phloroacetophenone monohydrate
  • CAS Number:207-556-5
  • Formula:C₈H₈O₄·1H₂O
  • MDL Number:MFCD00149091
  • Molecular Weight:186.16 g/mol
  • Boiling Pt:394 °C (1013 hPa)
  • Melting Pt:216…218 °C
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..