Order Entry
Italy
Orders LinkContactUsLinkComponent
Plugs for test tubes
16 mm, Red
Codice catalogo 212-0520
Nome del fornitore:   BOETTGER PAUL
Plugs for test tubes
Codice catalogo 212-0520
Nome del fornitore:  BOETTGER PAUL
Plugs for test tubes
Plugs for test tubes
16 mm, Red
Price
23,20 €
Log in for availability
1 * 1.000 pezzi
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
  • Not autoclavable
  • Easy handling with a high, ribbed grip
  • Easy to open by tilting 'one-hand operation'
Specifications
  • Colour:
    Red
  • Seq:
    08
  • Size:
    16 mm