Order Entry
Italy
Orders LinkContactUsLinkComponent
Gas sampling bags, Tedlar®

Gas sampling bags, Tedlar®

Gas sampling bags, 2-in-1 PP valve, 1 L, 178×178 mm
Catalog # 24633.
Supplier:   Merck
Gas sampling bags, Tedlar®
Catalog # 24633.
Supplier:  Merck
Gas sampling bags, Tedlar®
Gas sampling bags, Tedlar®

Gas sampling bags, 2-in-1 PP valve, 1 L, 178×178 mm

Price
184,00 €
Log in for availability
1 * 10 pezzi
Product Details & Documents

The unique valve and seam-sealing process ensure that these bags are sturdy and virtually leak-proof, even under the most demanding sampling and shipping conditions.

  • Valve consistently achieves a positive open or closed setting
  • 2 mil DuPont Tedlar material is used for superior inertness and impermeability
  • Patented polypropylene 2-in-1 valve has a unique push-pull function which allows the operating of bag’s valve effectively with one hand, while preparing a connection or withdrawing an aliquot with the other hand
Specifications
  • Capacity:
    1 L
  • Description:
    Gas sampling bags, 2-in-1 PP valve
  • Seq:
    01
  • W×L:
    178×178 mm