Order Entry
Italy
Orders LinkContactUsLinkComponent
PCR racks

PCR racks

96-Well PCR rack, Green
Catalog # RATI6114103
Supplier:   RATIOLAB
PCR racks
Catalog # RATI6114103
Supplier:  RATIOLAB
PCR racks
PCR racks

96-Well PCR rack, Green

Price
17,80 €
Log in for availability
1 * 10 pezzi
Product Details & Documents

Racks made of PP and are characterised by their very thin but uniform walls.

  • 8×12 positions, micro test plate format
  • Autoclavable
  • Available in different colors for the convenient identification of different types of samples
  • Transparent lids available in two different heights (Lids must be ordered seperately)

Work and storage rack for 0,2 ml and 0,5 ml PCR tubes, PCR strips, and PCR segments.

Specifications
  • Colour:
    Green
  • Description:
    96-Well PCR rack
  • Seq:
    03
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Lid, transparent, height 14 mm
    Lid, transparent, height 14 mm
    RATI6114110
    Log in for availability
    1 * 10 pezzi - 21,60 €

Product Information

  • Catalog #
    Availability
    Price
  • Lid, transparent, height 28 mm
    Lid, transparent, height 28 mm
    RATI6114111
    Log in for availability
    1 * 10 pezzi - 24,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.