Order Entry
Italy
Orders LinkContactUsLinkComponent
Cyclohexane 99.5%, extra pure

Cyclohexane 99.5%, extra pure

Glass bottle, 2,5 L
Catalog # ACRO210570025
Irritant
Health
Environment
Flammable
Cyclohexane 99.5%, extra pure
Catalog # ACRO210570025
CAS Number:  110-82-7
Cyclohexane 99.5%, extra pure
Cyclohexane 99.5%, extra pure

Glass bottle, 2,5 L

CAS Number:  110-82-7
Price
139,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (760 mmHg)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications