Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Cyclohexane 99.5%, extra pure

Glass bottle, 1 L
Catalog # ACRO210570010
Irritant
Health
Environment
Flammable
Cyclohexane 99.5%, extra pure
Catalog # ACRO210570010
CAS Number:  110-82-7
Cyclohexane 99.5%, extra pure
Cyclohexane 99.5%, extra pure

Glass bottle, 1 L

CAS Number:  110-82-7
Price
72,20 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (760 mmHg)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.