Order Entry
Italy
Orders LinkContactUsLinkComponent
Silver nitrate 99.5% for analysis

Silver nitrate 99.5% for analysis

Glass bottle, 25 g
Catalog # ACRO419360250
Oxidizing
Corrosive
Environment
Silver nitrate 99.5% for analysis
Catalog # ACRO419360250
CAS Number:  7761-88-8
Silver nitrate 99.5% for analysis
Silver nitrate 99.5% for analysis

Glass bottle, 25 g

CAS Number:  7761-88-8
Price
135,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:212 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration Argentometric:
    ≥99.4 %
  • Chloride (Cl):
    ≤5 ppm
  • Cadmium (Cd):
    ≤1 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Chromium (Cr):
    ≤0.2 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.4 ppm
  • Lead (Pb):
    ≤0.2 ppm
  • Manganese (Mn):
    ≤0.1 ppm
  • Nickel (Ni):
    ≤0.2 ppm
  • Palladium (Pd):
    ≤0.1 ppm
  • Sulfur (S):
    ≤20 ppm
  • Zinc (Zn):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications