2-Naphthaldehyde 98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: β-Naphthaldehyde
- CAS Number:66-99-9
- Formula:C₁₁H₈O
- MDL Number:MFCD00004094
- Molecular Weight:156.18 g/mol
- Boiling Pt:160 °C (19 mmHg)
- Melting Pt:58…62 °C
- Density:1,0775 g/cm³ (99 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-640-2
- UN:3335
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Off-white to yellow to light beige Powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:58 to 62 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



