Order Entry
Italy
Orders LinkContactUsLinkComponent
Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Sulfo-NHS-LC-biotin, 1 mg, 8
Catalog # 786-084
Supplier:   G-Biosciences
Biotin reagents, OneQuant™ HOOK™
Catalog # 786-084
Supplier:  G-Biosciences
CAS Number:  
Biotin reagents, OneQuant™ HOOK™
Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Sulfo-NHS-LC-biotin, 1 mg, 8
CAS Number:  
Price
260,00 €
Log in for availability
1 * 8 pezzi
Product Details & Documents
The OneQuant™ format prevents loss of reagent due to repeated weighing as each vial contains only 1 - 2 mg HOOK™ Biotin Reagent.
Specifications
Specifications
  • Pk:
    8
  • Description:
    OneQuant™ HOOK™ Sulfo-NHS-LC-biotin, 1 mg
Specifications