Accessories for Cryo Boxes
Dividers, PP, natural, length 121 mm, height 27 mm
Catalog # 211-0349
Supplier: GLW
Jump to:
Product Details & Documents
Documents
GLWAZ25_TDS_EN
Datasheet
Specifications
- Description:Dividers, PP, natural, length 121 mm, height 27 mm
- Seq:6
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



