Ethephon D4 (2-Chloroethyl-1,1,2,2-D4) 100 ng/µl 1 * 1 mL
Catalog # EHERXA13230100AC
Supplier: EHRENSTORFER
Ethephon D4 (2-Chloroethyl-1,1,2,2-D4) 100 ng/µl 1 * 1 mL
Catalog # EHERXA13230100AC
Supplier: EHRENSTORFER
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
UV/Visible standards, Supelco®
Supplier: Merck