Spouts for jerricans
Rigid spout, S60/61
Catalog # 590-0848
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
For simple and safe pouring of fluids from containers.
- Plastic
- Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Thread:S60/61
- Description:Rigid spout



