Ethanol, GPR RECTAPUR®
Plastic bottle, Duty paid, 1 L
: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
: Ethyl alcohol
- :64-17-5
- :C₂H₅OH
- :MFCD00003568
- :46.07 g/mol
- :78.3 °C (1013 hPa)
- :–117 °C
- :0.7895 g/cm³ (20 °C)
- :12 °C (closed cup)
- :Ambient
- :200-578-6
- :1170
- :3,II
- :01-2119457610-43
Documents
1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
20824_ETHANOL 96 % VOL GPR RECTAPUR_BSE_REV0
BSE/TSE Certificates
P20_VWRC20824.296_SPEC_EN.pdf P20_VWRC20824.321_SPEC_EN.pdf P20_VWRC20824.365_SPEC_EN.pdf P20_VWRC20824.467_SPEC_EN.pdf P20_VWRC20824.558_SPEC_EN.pdf
Product specification
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
- Assay (on anhydrous substance):Min. 99.8 %
- Assay (V/V):96.0 - 96.3 %
- Acidity:Max. 0.0002 meq/g
- Aldehydes (as CH3CHO):Max. 15 ppm
- Evaporation residue:Max. 20 ppm
- Pack type:Plastic bottle
- Pk:1 L
- Duty status:Duty paid



