Ethanol, GPR RECTAPUR®
Duty paid, 25 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.8 %
- Assay (V/V):96.0 - 96.3 %
- Acidity:Max. 0.0002 meq/g
- Aldehydes (as CH3CHO):Max. 15 ppm
- Evaporation residue:Max. 20 ppm
Specifications
- Pk:25 L
- Duty status:Duty paid



