Double bossheads
Double bosshead, safety screw type 'heart' M8, Stainless steel 18/10, 16,5 mm
Catalog # BOCH5354
Supplier: BOCHEM
Jump to:
Product Details & Documents
Double bossheads, either made of stainless steel or aluminium.
- Angle: 180 + 90°
- Different screw types available
Specifications
- Description:Double bosshead, safety screw type 'heart' M8
- Material:Stainless steel 18/10
- For rod Ø:16,5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



