Order Entry
Italy
Orders LinkContactUsLinkComponent
Stands for MSC-Advantage

Stands for MSC-Advantage

Fixed height floor stand, 750 mm working height, for 1,8 m cabinet
Catalog # 135-6833
Supplier:   Thermo Scientific
Stands for MSC-Advantage
Catalog # 135-6833
Supplier:  Thermo Scientific
Stands for MSC-Advantage
Stands for MSC-Advantage

Fixed height floor stand, 750 mm working height, for 1,8 m cabinet

Price
1 080,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Fixed height floor stand, 750 mm working height, for 1,8 m cabinet
  • Seq:
    03