Accessories for DOSE IT
Aspiration tube, stainless steel, one end dented, length: 35 cm, 6 mm
Catalog # 710-1039
Supplier: INTEGRA Biosciences
Jump to:
Product Details & Documents
Specifications
- Description:Aspiration tube, stainless steel, one end dented, length: 35 cm
- Ø int.:6 mm



