Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for DOSE IT

Accessories for DOSE IT

Silicone tubing, autoclavable, length: 25 m (bulk roll), 6 mm
Catalog # 710-1032
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1032
Accessories for DOSE IT
Accessories for DOSE IT

Silicone tubing, autoclavable, length: 25 m (bulk roll), 6 mm

Price
188,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Documents

Specifications
  • Description:
    Silicone tubing, autoclavable, length: 25 m (bulk roll)
  • Ø int.:
    6 mm