Ion selective electrode system, AmmoLyt® Plus
Ion selective electrode set, AmmoLyt® Plus, 1 - 1000 mg/l (NH₄-N)
1 - 1290 mg/l (NH₄+), 0...+40 °C
1 - 1290 mg/l (NH₄+), 0...+40 °C
Catalog # WTWO107071
Supplier: WTW ONLINE
Jump to:
Product Details & Documents
This system – consisting of a robust, digital armature with ammonium replacement electrode and ammonium reference electrode – allows ammonium measurement directly in the medium, without sample preparation. The system is designed for connection to the IQ SENSOR NET multiparameter measuring system.
- Calibration-free, long term stable
- With integrated temperature sensor
- Short response time
- With screw/plug connector for connection of the SACIQ sensor connection cable
: Supplied with unequipped sensor AmmoLyt® Plus 700 IQ, reference electrode VARiON® Ref, VARiON® Plus NH4 (ammonium electrode), special socket wrench, protective hood VARiON® 700 IQ-SK, Potassium chloride solution for storing the reference electrode and operating manual.
Specifications
- Concentration:1 - 1000 mg/l (NH₄-N)
1 - 1290 mg/l (NH₄+) - Description:Ion selective electrode set, AmmoLyt® Plus
- Temp.:0...+40 °C