Water
50 kg
Supplier: ..
Water
Catalog # SILM04284.K050
Supplier: ..
CAS Number: 7732-18-5
Jump to:
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1 g/cm³ (4 °C)
- Storage Temperature:Ambient
- EINECS:231-791-2
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:50 kg
- Cat. No.:SILM04284.K050
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



