Order Entry
Italy
ContactUsLinkComponent
Fully automatic, microprocessor-controlled benchtop laboratory autoclave with CH-plugChamberdimension (xL) 380 x 690 mm Volume: 85 liters 1 * 1 items
Catalog # 481-2056
Supplier:  TUTTNAUER
Fully automatic, microprocessor-controlled benchtop laboratory autoclave with CH-plugChamberdimension (xL) 380 x 690 mm Volume: 85 liters 1 * 1 items
Catalog # 481-2056
Supplier:  TUTTNAUER