CORNING CO2 REAGENT 6 X 200ML 1 * 1 Each
Catalog # REGERR021
Supplier: REAGECON
Frequently Bought Together
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, Bioreagent
Supplier: Thermo Fisher Scientific
Carbon disulphide ≥99.9% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur. for IR spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific