Order Entry
Italy
Orders LinkContactUsLinkComponent
Spring tubing clips, SAMCO

Spring tubing clips, SAMCO

12 mm
Catalog # MURRL150/02
Supplier:   MURRAY S
Spring tubing clips, SAMCO
Catalog # MURRL150/02
Supplier:  MURRAY S
Spring tubing clips, SAMCO

12 mm

Price
415,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

These spring tubing clips are made from chemical and stain resistant acetal copolymer and stainless steel.

  • Operates with parallel action to provide optimum closure of rubber and flexible PVC tubing
  • Size 8 accepts tubing up to 8 mm outside diameter
  • Size 12 accepts tubing up to 12 mm outside diameter
Specifications
  • Spanning width:
    12 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.