Order Entry
Italy
Orders LinkContactUsLinkComponent
Spring tubing clips, SAMCO

Spring tubing clips, SAMCO

8 mm
Catalog # MURRL150/01
Supplier:   MURRAY S
Spring tubing clips, SAMCO
Catalog # MURRL150/01
Supplier:  MURRAY S
Spring tubing clips, SAMCO

8 mm

Price
368,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

These spring tubing clips are made from chemical and stain resistant acetal copolymer and stainless steel.

  • Operates with parallel action to provide optimum closure of rubber and flexible PVC tubing
  • Size 8 accepts tubing up to 8 mm outside diameter
  • Size 12 accepts tubing up to 12 mm outside diameter
Specifications
  • Spanning width:
    8 mm