Watch glasses
Watch glass, glass, with ground edges, 50 mm
Catalog # MURRG850/01
Supplier: MURRAY S
Jump to:
Product Details & Documents
Made of glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With ground edges for covering beakers.
Specifications
- Ø:50 mm
- Descrizione:Watch glass, glass, with ground edges
- Seq:01