Overhead stirrer tools, anchor stirrers
Anchor stirrer, DURAN®, 60 LF, 1000 ml, 10 mm
LENZ06320670
Anchor stirrers with GL thread connection.
- Made of precision calibrated glass tube
- With ground and polished bearing surface
- Suitable for flange vessels
- Description:Anchor stirrer, DURAN®, 60 LF
- Shaft Ø:10 mm
- For container:1000 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



