Labels for bottles
Labels for Methylene Cloride, Black, 130×35 mm
Catalog # KART5018
Supplier: KARTELL
Jump to:
Product Details & Documents
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles.
- Complete with risk assessment numbers
- Recommended by the NFPA (National Fire Protection Association)
Specifications
- Colour:Black
- Description:Labels for Methylene Cloride
- Dimensions:130×35 mm