Dry box gloves
Seamless gloves for all Labconco glove boxes, 8,5, Black, 0,015 gauge
LBCP5005500
: LABCONCO
- Glove size:8,5
- Colour:Black
- Thickness:0,015 gauge
- Model:Seamless gloves for all Labconco glove boxes
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
#: 20821.365
: VWR Chemicals



