Order Entry
Italy
Orders LinkContactUsLinkComponent
Sleeves for conical ground joints
Sleeves for conical ground joints
19/38
Codice catalogo COWI019.019
Nome del fornitore:   COWIE
Sleeves for conical ground joints
Codice catalogo COWI019.019
Nome del fornitore:  COWIE
Sleeves for conical ground joints
Sleeves for conical ground joints
19/38
Price
2,90 €
Log in for availability
1 * 1 pezzi
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Thin wall joint sleeves, made of PFTE, are inserted between standard taper ground glass joints to eliminate the use of grease and prevent sticking.
  • Sleeves have a wall thickness of about 0.05mm are inert
  • Usable to 280˚C
  • Withstand low to medium vacuum
Specifications
  • For NS:
    19/38
  • Seq:
    07