Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Hexene ≥98%

1-Hexene ≥98%

bottiglia in vetro, 100 mL
Catalog # B20271.AE
Flammable
Health
1-Hexene ≥98%
Catalog # B20271.AE
CAS Number:  592-41-6
1-Hexene ≥98%
1-Hexene ≥98%

bottiglia in vetro, 100 mL

CAS Number:  592-41-6
Price
24,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 98% CAS No.: 592-41-6 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -25°(-13°F) Refractive Index: 1.3880 Odour: Mild Melting Point: -140° Boiling Point: 62-64° Beilstein Reference: 1209240 UN No.: 2370 MDL No.: MFCD00009505

  • CAS Number:592-41-6
  • Formula:C₆H₁₂
  • MDL Number:MFCD00009505
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:62…64 °C
  • Melting Pt:–140 °C
  • Density:0,673
  • Flash Pt:–25 °C (–13 °F)
  • Storage Temperature:Ambiente
  • EINECS:209-753-1
  • UN:2370
  • ADR:3,II
Specifications

Specifications

  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.