Order Entry
Italy
Orders LinkContactUsLinkComponent
7-Hydroxyflavone ≥98%

7-Hydroxyflavone ≥98%

Glass bottle for solids, 5 g
Catalog # A18538.06
Irritant
7-Hydroxyflavone ≥98%
Catalog # A18538.06
CAS Number:  6665-86-7
7-Hydroxyflavone ≥98%
7-Hydroxyflavone ≥98%

Glass bottle for solids, 5 g

CAS Number:  6665-86-7
Price
173,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 6665-86-7 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 245-247° Beilstein Reference: 194089

Synonyms:  7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
  • CAS Number:6665-86-7
  • Formula:C₁₅H₁₀O₃
  • MDL Number:MFCD00006835
  • Molecular Weight:238.24 g/mol
  • Boiling Pt:395 °C (1013 hPa)
  • Melting Pt:245…247 °C
  • Storage Temperature:Ambient
  • EINECS:229-705-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications