Order Entry
Italy
Orders LinkContactUsLinkComponent
7-Hydroxyflavone ≥98%

7-Hydroxyflavone ≥98%

Glass bottle for solids, 1 g
Catalog # A18538.03
Irritant
7-Hydroxyflavone ≥98%
Catalog # A18538.03
CAS Number:  6665-86-7
7-Hydroxyflavone ≥98%
7-Hydroxyflavone ≥98%

Glass bottle for solids, 1 g

CAS Number:  6665-86-7
Price
53,50 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 6665-86-7 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 245-247° Beilstein Reference: 194089

Synonyms:  7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
  • CAS Number:6665-86-7
  • Formula:C₁₅H₁₀O₃
  • MDL Number:MFCD00006835
  • Molecular Weight:238.24 g/mol
  • Boiling Pt:395 °C (1013 hPa)
  • Melting Pt:245…247 °C
  • Storage Temperature:Ambiente
  • EINECS:229-705-3
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.