Order Entry
Italy
Orders LinkContactUsLinkComponent
Adipic acid ≥99%

Adipic acid ≥99%

Glass bottle for solids, 5.000 g
Catalog # A13705.0I
Irritant
Adipic acid ≥99%
Catalog # A13705.0I
CAS Number:  124-04-9
Adipic acid ≥99%
Adipic acid ≥99%

Glass bottle for solids, 5.000 g

CAS Number:  124-04-9
Price
108,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 124-04-9 Molecular Formula: HOOC(CH2)4COOH Formula Weight: 146.14 Flash Point: 196°(384°F) Odour: Odourless pH: 2.7 Melting Point: 151-155° Boiling Point: ca 205°/3mm Beilstein Reference: 1209788 Merck Reference: 14,162 MDL No.: MFCD00004420

Synonyms:  Hexanedioc acid, 1,6-Hexanedioic acid
  • CAS Number:124-04-9
  • Formula:HOOC(CH₂)₄COOH
  • MDL Number:MFCD00004420
  • Molecular Weight:146.14 g/mol
  • Merck Index:14,162
  • Boiling Pt:∼205 °C (3 mmHg)
  • Melting Pt:151…155 °C
  • Density:1.360
  • Flash Pt:196 °C (384 °F)
  • Storage Temperature:Ambient
  • EINECS:204-673-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.