Order Entry
Italy
Orders LinkContactUsLinkComponent
Adipic acid ≥99%
Glass bottle for solids, 25.000 g
  A13705.0G
Adipic acid ≥99%
  A13705.0G
 :  124-04-9
Adipic acid ≥99%
Adipic acid ≥99%
Glass bottle for solids, 25.000 g
 :  124-04-9
415,00 €
1 * 25.000 g
Purity: 99% CAS No.: 124-04-9 Molecular Formula: HOOC(CH2)4COOH Formula Weight: 146.14 Flash Point: 196°(384°F) Odour: Odourless pH: 2.7 Melting Point: 151-155° Boiling Point: ca 205°/3mm Beilstein Reference: 1209788 Merck Reference: 14,162 MDL No.: MFCD00004420
 :  Hexanedioc acid, 1,6-Hexanedioic acid
  •  :124-04-9
  •  :HOOC(CH₂)₄COOH
  •  :MFCD00004420
  •  :146.14 g/mol
  •  :14,162
  •  :∼205 °C (3 mmHg)
  •  :151…155 °C
  •  :1.360
  •  :196 °C (384 °F)
  •  :Ambient
  •  :204-673-3
  • Pack type:
    Glass bottle for solids
  • Pk:
    25.000 g