Order Entry
Italy
Orders LinkContactUsLinkComponent
3-Mercaptopropionic acid ≥99%

3-Mercaptopropionic acid ≥99%

Glass bottle for solids, 100 g
Catalog # A13261.22
Toxic
Corrosive
3-Mercaptopropionic acid ≥99%
Catalog # A13261.22
CAS Number:  107-96-0
3-Mercaptopropionic acid ≥99%
3-Mercaptopropionic acid ≥99%

Glass bottle for solids, 100 g

CAS Number:  107-96-0
Price
32,70 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 107-96-0 Molecular Formula: C3H6O2S Formula Weight: 106.14 Flash Point: 93°(199°F) Refractive Index: 1.4920 Melting Point: 17-19° Boiling Point: 110-111°/15mm Beilstein Reference: 773807 UN No.: 2922 MDL No.: MFCD00004897

  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:110…111 °C (15 mmHg)
  • Melting Pt:17…19 °C
  • Density:1.220
  • Flash Pt:93 °C (199 °F)
  • Storage Temperature:Ambient
  • EINECS:203-537-0
  • UN:2922
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications