Order Entry
Italy
Orders LinkContactUsLinkComponent
DL-Proline ≥99%

DL-Proline ≥99%

Glass bottle for solids, 100 g
Catalog # A13137.22
DL-Proline ≥99%
Catalog # A13137.22
CAS Number:  609-36-9
DL-Proline ≥99%
DL-Proline ≥99%

Glass bottle for solids, 100 g

CAS Number:  609-36-9
Price
472,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 609-36-9 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: ca 209° dec. Beilstein Reference: 80809 Merck Reference: 14,7780 MDL No.: MFCD00005250

Synonyms:  (±)-Proline, H-DL-Pro-OH
  • CAS Number:609-36-9
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00005250
  • Molecular Weight:115.13 g/mol
  • Merck Index:14,7780
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:∼ 209 °C (dec.)
  • Density:1.38 g/cm³ (22.9 °C)
  • Storage Temperature:Ambient
  • EINECS:210-189-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications