Order Entry
Italy
Orders LinkContactUsLinkComponent
 

DL-Proline ≥99%

Glass bottle for solids, 25 g
Catalog # A13137.14
DL-Proline ≥99%
Catalog # A13137.14
CAS Number:  609-36-9
DL-Proline ≥99%
DL-Proline ≥99%

Glass bottle for solids, 25 g

CAS Number:  609-36-9
Price
161,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 609-36-9 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: ca 209° dec. Beilstein Reference: 80809 Merck Reference: 14,7780 MDL No.: MFCD00005250

Synonyms:  (±)-Proline, H-DL-Pro-OH
  • CAS Number:609-36-9
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00005250
  • Molecular Weight:115.13 g/mol
  • Merck Index:14,7780
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:∼ 209 °C (dec.)
  • Density:1.38 g/cm³ (22.9 °C)
  • Storage Temperature:Ambient
  • EINECS:210-189-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.