Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2-Naphthaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # B25157.14
Toxic
Irritant
2-Naphthaldehyde ≥98%
Catalog # B25157.14
CAS Number:  66-99-9
2-Naphthaldehyde ≥98%
2-Naphthaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  66-99-9
Price
111,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 66-99-9 Molecular Formula: C11H8O Formula Weight: 156.18 Melting Point: 59-62° Boiling Point: 160°/19mm MDL No.: MFCD00004094

Synonyms:  β-Naphthaldehyde
  • CAS Number:66-99-9
  • Formula:C₁₁H₈O
  • MDL Number:MFCD00004094
  • Molecular Weight:156.18 g/mol
  • Boiling Pt:160 °C (19 mmHg)
  • Melting Pt:59…62 °C
  • Density:1.0775 g/cm³ (99 °C)
  • Storage Temperature:Ambient
  • EINECS:200-640-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.