2-Naphthaldehyde ≥98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 66-99-9 Molecular Formula: C11H8O Formula Weight: 156.18 Melting Point: 59-62° Boiling Point: 160°/19mm MDL No.: MFCD00004094
Synonyms: β-Naphthaldehyde
- CAS Number:66-99-9
- Formula:C₁₁H₈O
- MDL Number:MFCD00004094
- Molecular Weight:156.18 g/mol
- Boiling Pt:160 °C (19 mmHg)
- Melting Pt:59…62 °C
- Density:1.0775 g/cm³ (99 °C)
- Storage Temperature:Ambient
- EINECS:200-640-2
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



